ENA238711401 Enamine

[2-(3-fluorophenoxy)ethyl](methyl)amine

  • MDL number MFCD06247666
  • Cas Number : 883540-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45194
Availability details for
Quantity
  • Availability
  • Add to Cart