ENA238711405 Enamine

1-(4-bromophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09941736
  • Cas Number : 138907-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44129
Availability details for
Quantity
  • Availability
  • Add to Cart