ENA238711410 Enamine

2-hydroxy-1-methyl-3-nitro-1,4-dihydroquinolin-4-one

  • MDL number MFCD03813459
  • Cas Number : 672895-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45209
Availability details for
Quantity
  • Availability
  • Add to Cart