ENA238711412 Enamine

2-chloro-5-(methylsulfanyl)benzamide

  • MDL number MFCD06384552
  • Cas Number : 548456-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45213
Availability details for
Quantity
  • Availability
  • Add to Cart