ENA238711423 Enamine

4-ethoxybutan-1-amine

  • MDL number MFCD09808783
  • Cas Number : 58114-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45238
Availability details for
Quantity
  • Availability
  • Add to Cart