ENA238711429 Enamine

2-(3-nitrophenyl)-1,3-thiazole

  • MDL number MFCD12197177
  • Cas Number : 105361-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238...240
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45260
Availability details for
Quantity
  • Availability
  • Add to Cart