ENA238711432 Enamine

methyl[2-(3-methylphenoxy)ethyl]amine hydrochloride

  • MDL number MFCD08727962
  • Cas Number : 1050509-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-45266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45266
Availability details for
Quantity
  • Availability
  • Add to Cart