ENA238711437 Enamine

7-chloro-5-ethyl-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD08898721
  • Cas Number : 167371-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45274
Availability details for
Quantity
  • Availability
  • Add to Cart