ENA238711443 Enamine

2-[2-(1-aminopropyl)phenoxy]-N-methylacetamide

  • MDL number MFCD09815360
  • Cas Number : 1016521-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45282
Availability details for
Quantity
  • Availability
  • Add to Cart