ENA238711448 Enamine

5-[(2-fluorophenyl)methyl]-4-methyl-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD12197183
  • Cas Number : 1181458-64-9
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45293
Availability details for
Quantity
  • Availability
  • Add to Cart