ENA238711451 Enamine

4-methyl-5-[(3-methylphenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12197363
  • Cas Number : 1181457-72-6
  • Molecular Weight : 254.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45299
Availability details for
Quantity
  • Availability
  • Add to Cart