ENA238711452 Enamine

5-[(3-chlorophenyl)methyl]-4-methyl-1,3-thiazol-2-amine

  • MDL number MFCD12096064
  • Cas Number : 1155998-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45301
Availability details for
Quantity
  • Availability
  • Add to Cart