ENA238711454 Enamine

1-[(3-aminophenyl)methyl]-1,2-dihydropyrimidin-2-one

  • MDL number MFCD09810076
  • Cas Number : 1016704-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45306
Availability details for
Quantity
  • Availability
  • Add to Cart