ENA238711455 Enamine

2-(3-aminophenoxy)-N-phenylacetamide

  • MDL number MFCD09812954
  • Cas Number : 92906-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120...122
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45308

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45308
Availability details for
Quantity
  • Availability
  • Add to Cart