ENA238711471 Enamine

4-amino-N-cyclohexyl-3-methylbenzamide

  • MDL number MFCD09045915
  • Cas Number : 926227-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45383
Availability details for
Quantity
  • Availability
  • Add to Cart