ENA238711472 Enamine

3-fluoro-4-(1H-1,2,4-triazol-1-yl)aniline

  • MDL number MFCD08699587
  • Cas Number : 181997-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45385
Availability details for
Quantity
  • Availability
  • Add to Cart