ENA238711479 Enamine

2-amino-1-(2,6-dimethylmorpholin-4-yl)-2-phenylethan-1-one hydrochloride

  • MDL number MFCD12197198
  • Cas Number : 1214808-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45401
Availability details for
Quantity
  • Availability
  • Add to Cart