ENA238711481 Enamine

2-amino-1-(4-methylpiperidin-1-yl)-2-phenylethan-1-one hydrochloride

  • MDL number MFCD12197199
  • Cas Number : 1214125-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45404
Availability details for
Quantity
  • Availability
  • Add to Cart