ENA238711484 Enamine

3-(1,3-benzothiazol-2-yl)propan-1-ol

  • MDL number MFCD09926855

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45411
Availability details for
Quantity
  • Availability
  • Add to Cart