ENA238711486 Enamine

(diphenyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD12197201
  • Cas Number : 56999-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45419
Availability details for
Quantity
  • Availability
  • Add to Cart