ENA238711490 Enamine

2-(morpholin-4-yl)-5-phenyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05742302
  • Cas Number : 724746-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 307...309
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45426

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45426
Availability details for
Quantity
  • Availability
  • Add to Cart