ENA238711492 Enamine

4-acetyl-5-methyl-3-propyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD12197202
  • Cas Number : 1181894-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45428
Availability details for
Quantity
  • Availability
  • Add to Cart