ENA238711493 Enamine

5-(3,5-dimethylpiperidine-1-carbonyl)-2-methylaniline

  • MDL number MFCD09737230
  • Cas Number : 954274-68-1
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45429
Availability details for
Quantity
  • Availability
  • Add to Cart