ENA238711496 Enamine

(3-bromophenyl)urea

  • MDL number MFCD00041317
  • Cas Number : 2989-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45433
Availability details for
Quantity
  • Availability
  • Add to Cart