ENA238711499 Enamine

3-ethanesulfonamidopropanoic acid

  • MDL number MFCD08444792
  • Cas Number : 923249-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45436

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45436
Availability details for
Quantity
  • Availability
  • Add to Cart