ENA238711504 Enamine

N-(1,3,4-thiadiazol-2-yl)piperidine-3-carboxamide

  • MDL number MFCD11128708
  • Cas Number : 1019324-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-45443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45443
Availability details for
Quantity
  • Availability
  • Add to Cart