ENA238711505 Enamine

2-amino-N-(4-bromophenyl)acetamide hydrochloride

  • MDL number MFCD12197204
  • Cas Number : 1181458-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45444
Availability details for
Quantity
  • Availability
  • Add to Cart