ENA238711525 Enamine

2-amino-5-phenylthiophene-3-carbonitrile

  • MDL number MFCD06803446
  • Cas Number : 60271-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45475
Availability details for
Quantity
  • Availability
  • Add to Cart