ENA238711530 Enamine

N-methyl-2-(methylamino)-N-(1-methylpiperidin-4-yl)acetamide dihydrochloride

  • MDL number MFCD12197217
  • Cas Number : 1181519-34-5
  • Molecular Weight : 272.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45481
Availability details for
Quantity
  • Availability
  • Add to Cart