ENA238711531 Enamine

5-(pyridin-2-yl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD05740063
  • Cas Number : 25468-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44158
Availability details for
Quantity
  • Availability
  • Add to Cart