ENA238711538 Enamine

2-(4-chlorophenyl)-2-methylpropan-1-amine

  • MDL number MFCD09903953
  • Cas Number : 42299-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45493
Availability details for
Quantity
  • Availability
  • Add to Cart