ENA238711541 Enamine

2-(2,5-dioxopyrrolidin-1-yl)ethanethioamide

  • MDL number MFCD09933518
  • Cas Number : 1016784-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45504
Availability details for
Quantity
  • Availability
  • Add to Cart