ENA238711544 Enamine

2-[(2-methylfuran-3-yl)formamido]acetic acid

  • MDL number MFCD09049417
  • Cas Number : 926226-44-0
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45508
Availability details for
Quantity
  • Availability
  • Add to Cart