ENA238711547 Enamine

N-(pyrimidin-2-yl)piperidine-4-carboxamide

  • MDL number MFCD09948244
  • Cas Number : 1016865-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45514

Safety Information

signalword Warning
pcodes P264,P280,P302+P352,P305+P351+P338,P332+P313,P337+P313,P362
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-45514
pictograms GHS07
hstatements H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart