ENA238711549 Enamine

2-cyclohexylpyrrolidine

  • MDL number MFCD01631841
  • Cas Number : 367281-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45516
Availability details for
Quantity
  • Availability
  • Add to Cart