ENA238711557 Enamine

2-{4-[(hydroxyimino)methyl]-3,5-dimethyl-1H-pyrazol-1-yl}acetic acid

  • Cas Number : 1181471-99-7
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45524
Availability details for
Quantity
  • Availability
  • Add to Cart