ENA238711559 Enamine

naphthalene-2-carbohydrazide

  • MDL number MFCD00016810
  • Cas Number : 39627-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44166

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-44166
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart