ENA238711560 Enamine

3-(1,3,4-oxadiazol-2-yl)pyridine

  • MDL number MFCD00816745
  • Cas Number : 65943-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44169
Availability details for
Quantity
  • Availability
  • Add to Cart