ENA238711562 Enamine

tert-butyl N-(2-aminocycloheptyl)carbamate

  • MDL number MFCD12197222
  • Cas Number : 1193390-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45539
Availability details for
Quantity
  • Availability
  • Add to Cart