ENA238711564 Enamine

1-ethyl-1H-1,3-benzodiazole-2-carbaldehyde

  • MDL number MFCD01051249
  • Cas Number : 34734-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45541
Availability details for
Quantity
  • Availability
  • Add to Cart