ENA238711568 Enamine

N-[(1-methyl-1H-pyrazol-4-yl)methyl]-2-(trifluoromethyl)aniline

  • MDL number MFCD04968275
  • Cas Number : 1006466-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45552
Availability details for
Quantity
  • Availability
  • Add to Cart