ENA238711575 Enamine

4-(2-methoxyethoxy)-2-methylaniline hydrochloride

  • MDL number MFCD09997933
  • Cas Number : 1185156-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45561
Availability details for
Quantity
  • Availability
  • Add to Cart