ENA238711580 Enamine

4-methyl-5-{[4-(trifluoromethoxy)phenyl]methyl}-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12197082
  • Cas Number : 1171936-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44172
Availability details for
Quantity
  • Availability
  • Add to Cart