ENA238711582 Enamine

2-[2-(1-aminopropyl)phenoxy]-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD09729346
  • Cas Number : 953731-69-6
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-{2-[2-oxo-2-(1-pyrrolidinyl)ethoxy]phenyl}-1-propanamine
Inchi Code 1S/C15H22N2O2/c1-2-13(16)12-7-3-4-8-14(12)19-11-15(18)17-9-5-6-10-17/h3-4,7-8,13H,2,5-6,9-11,16H2,1H3
Inchi Key XSHNCPFJHROPFL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45571

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-45571
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart