ENA238711588 Enamine

1-(3,4-dichlorophenyl)ethan-1-ol

  • MDL number MFCD00021865
  • Cas Number : 1475-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45582

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-45582
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart