ENA238711589 Enamine

(4-butylcyclohexyl)methanol

  • MDL number MFCD11646255
  • Cas Number : 376642-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-45583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45583
Availability details for
Quantity
  • Availability
  • Add to Cart