ENA238711594 Enamine

(isocyanatomethyl)cyclopentane

  • MDL number MFCD09943483
  • Cas Number : 42079-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48600

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48600
Availability details for
Quantity
  • Availability
  • Add to Cart