ENA238711598 Enamine

4-amino-2-chloro-N-(2-methylbutan-2-yl)benzamide hydrochloride

  • MDL number MFCD12197230
  • Cas Number : 1193388-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48604

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48604
Availability details for
Quantity
  • Availability
  • Add to Cart