ENA238711602 Enamine

N-butyl-N-methylpiperidine-4-carboxamide hydrochloride

  • MDL number MFCD12197367
  • Cas Number : 1193389-21-7
  • Molecular Weight : 234.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48608
Availability details for
Quantity
  • Availability
  • Add to Cart