ENA238711604 Enamine

4-cyano-N-[3-(dimethylamino)propyl]benzene-1-sulfonamide hydrochloride

  • MDL number MFCD12197231
  • Cas Number : 1193390-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48611
Availability details for
Quantity
  • Availability
  • Add to Cart